Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPAN Diagnostics UTM™ Viral Specimen Collection Kit
SDP

The Viral Specimen Collection Kit is intended for the collection, transport and preservation of clinical specimens containing viruses for viral molecular diagnostic testing, including COVID-19, chlamydia, mycoplasma or ureaplasma organisms

Supplier:  COPAN Diagnostics 3C057N

 View more versions of this product

Catalog No. 23-600-989



Explore available promotions
Add to Cart

Description

Description

  • Room temperature stable
  • Contains antibiotics to inhibit bacterial and fungal flora in patient samples
  • Maintains viability of organisms for up to 48 hours
  • Provided in labeled screw-cap tubes designed for transport of the clinical sample
Specifications

Specifications

Flocked Collection Kit
Sterile
3 mL
Refrigerate at 2°C to 8°C or kept on wet ice following collection and while in transit
16 x 100 mm
UTM
Minitip
50 Pk.
Nylon
Plastic
Safety and Handling

Safety and Handling

ShelfLife : 18 Months

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Intended only for use with viral molecular tests.