Learn More
SA Scientific Urine hCG, Serum/Urine hCG, and Ultra™ hCG Serum/Urine Tests
Results in as little as four minutes
Supplier: SA Scientific COL087715
This item is not returnable.
View return policy
Description
- CLIA Complexity: Waived for Urine Only
- CPT Codes: 81025 (Waived), 84703 (Non-waived)
- Available with choice of sensitivity—10 or 20mIU/mL in Ultra Serum/Urine or 25mIU/mL in Serum/Urine and urine only tests
- Visual, one-step test for the qualitative determination of human chorionic gonadotropin (hCG) in serum or urine to aid in early detection of pregnancy
- Results in four to eight minutes
- Utilizes combination of monoclonal and polyclonal antibody reagent to selectively detect elevated levels of hCG
- Positive specimens react with the specific antibody-hCG colored conjugate and form a colored line in the S (specimen) area; absence of this colored line suggests a negative result
- A colored line in the C (control) area will always appear regardless of the presence or absence of hCG
- Addition of LH (300mIU/mL), FSH (1000mIU/mL) and TSH (1000μIU/mL) to negative (0mIU/mL hCG) or positive (25mIU/mL hCG) serum/urine samples showed no cross-reactivity
Specifications
84703 | |
hCG | |
Ultra Serum Control Set | |
3 x 5 mL |
Non-waived | |
Serum | |
Negative, low positive and high positive control | |
4 to 8 min. |