Learn More
Electron Microscopy Sciences Uranyl Zinc Acetate 5 GR
Supplier: Electron Microscopy Sciences 22610
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
CAS #10138-94-0 ZnUO₄(CH₃COO)₂ F.W. 571.59
Used as a laboratory reagent in the determination of sodium concentrations of solutions.