Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Celestron™ UpClose G2 Binoculars
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
High Quality Binoculars at an affordable price
Supplier: Celestron™ 71258
Includes: Soft Carrying Case, Binocular Straps
Description
Features:
- Multi-coated optics increase light transmission resulting in brighter and sharper images with high contrast levels
- Stylish rubber covered aluminum body protects from rough handling
- Ergonomic thumb indents and finger ridges for extra non-slip comfort and ease of use
- Water resistant
- Soft carrying case
- Binocular straps
Ideal For:
- Bird Watching
- Sporting Events
- Nature & Wildlife
- Hunting
- Travel & ventures
- Concerts & live events
- Boating
Warranty and Services
Limited Lifetime
Specifications
Roof Prism | |
1.75 lb. | |
20x | |
Binoculars |
Strap and carrying case | |
No | |
794 g |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction