Learn More
Cytiva Untagged Column Package
Combines cation and anion exchange with size exclusion chromatography for purification of most untagged or native proteins
Supplier: Cytiva 29497631
Description
- Package contains HiTrap™ Capto Q ImpRes (1 x 1 mL) and Capto SP ImpRes (1 x 1 mL) Ion Exchange (IEX) columns for effective first-step purification
- Obtain final high purity with HiPrep™ 16/60 Sephacryl S-200 HR: a Size Exclusion Chromatography (SEC) column with a broad fractionation range of Mr 5000 to ∼ 250 000 for globular proteins
- Includes instructions with guidance and tips to quickly get started
Specifications
Untagged Column | |
1 Kit | |
4°C to 30°C |
Purification of Most Untagged or Native Proteins | |
HiTrap&Trade; Capto Q ImpRes (1 x 1 mL) Column, Capto SP ImpRes (1 x 1 mL) Ion Exchange (IEX) Column, and HiPrep™ 16/60 Sephacryl S-200 HR Size Exclusion Chromatography Column |