Learn More
BURKLE INC Uno sampler, V4A/PTFE, 85 cm
Supplier: BURKLE INC 53161085
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Uno sampler made of stainless steel AISI 316L (1.4404)/PTFE with a closed sample chamber for point sampling from a predetermined sampling depth. Burkle zone samplers are used for taking samples of bulk goods of any kind. Various bulk goods can be collected with the lances, from the finest powder to large grains such as corn or nuts.The zone samplers are available in different lengths. 55 cm (21.65 in.) especially suitable for sacks, 85 cm (33.46 in.) long (especially for barrels, tubs, mixed containers) and 150 cm (59.06 in.) as well as 250 cm (98.43 in.) (only Jumbo sampler) for big bags, silos, tanks. Safe and reliable sampling is made even easier with useful special accessories such as sealing sleeves, Quali-Paper, cleaning brushes, emptying devices and close-it control seals.The zone samplers are available in different models for different sampling tasks.Other versions of zone samplers: Multi Sampler and All-layer-sampler
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.