Learn More
Malvern Panalytical Universal 'Dip' Cell Kit, Compatible with all Nano Series Systems that measure Zeta Potential
Supplier: Malvern Panalytical ZEN1002
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Universal 'dip' cell kit. For use with samples in aqueous and non-aqueous, i.e. non-polar dispersants such as hydrocarbons. Compatible with PCS1115 cuvettes. Compatible with all Nano series systems that measure zeta potential.