Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bollé Safety™ Unifocal Photochromic Trivex Rx Lens
Prescription lens option for safety glasses.
$214.50 - $415.50
Specifications
Lens Coating | Uncoated |
---|---|
Lens Type | Single Vision Photochromatic |
Product Type | Prescription Lens |
Disposable | Reusable |
Lens Material | Plastic |
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
18-072-194
|
Bollé Safety™
PXLUNIST02 |
Brown |
Each for $214.50
|
|
|||||
18-072-195
|
Bollé Safety™
PXLUNIST03 |
Gray |
Each for $415.50
|
|
|||||
Description
- Lenses made of plastic
- Available in multiple colors
Specifications
Uncoated | |
Prescription Lens | |
Plastic |
Single Vision Photochromatic | |
Reusable | |
Not Autoclavable |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title