Learn More
Glass Expansion UniFit with 2.0mm OD x 0.86mm
Supplier: Glass Expansion UFT286
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
UniFit with 2.0mm OD x 0.86mm ID x 480mm long tube (PKT 3). Replacement pack of 3 spray chamber drain lines. Typically used with IsoMist XR spray chambers.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001