Learn More
Glass Expansion UniFit-EzyFit Sample Tube (PKT 6)
Supplier: Glass Expansion UFTEZT1650
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
UniFit-EzyFit Sample Tube (PKT 6). A replacement 1.6mm (1/16”) OD and 0.50mm ID capillary with a UniFit connector on one end and an EzyFit connector on the other end. Typically used with Glass Expansion’s Trident IS Kit for HF P/N NC2046964, 60-808-1150. This fitting connects the mixed sample and internal standard from the mixing T to a Glass Expansion U-Series or Direct Connect “DC” nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.