Learn More
TARGETMOL CHEMICALS INC UNC10217938A HCL 5MG
Supplier: TARGETMOL CHEMICALS INC T13254L
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. UNC10217938A HCl (UNC7938 HCl) is a 3-deazapteridine analog. UNC10217938A HCl exhibits strong oligonucleotide enhancing effects. UNC10217938A HCl can modulate their intracellular trafficking and release from endosomes leading to enhance oligonucleotides effects. UNC10217938A HCl also enhances the effects of siRNA and antisense oligonucleotides.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.