Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ UNC 0642
Potent and selective G9a and GLP inhibitor
$387.44 - $1605.44
Specifications
Purity | >99% |
---|---|
Content And Storage | -20°C |
Product Type | UNC 0642 |
Molecular Weight (g/mol) | 546.7 |
Inhibitors | UNC 0642 |
Description
Potent and selective G9a and GLP histone lysine methyltransferase inhibitor (IC50 < 2.5 nM). Exhibits >2,000-fold selectivity for G9a and GLP over PRC2-EZH2 and >20,000-fold selectivity over other methyltransferases. Reduces H3K9 dimethylation levels in MDA-MB-231 cells (IC50 = 110 nM). Displays modest brain penetration in vivo.Specifications
>99% | |
UNC 0642 | |
UNC 0642 |
-20°C | |
546.7 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title