Learn More
World Precision Instrument UltraDose Germicidal Solution
Supplier: World Precision Instrument WPI036
This EPA approved super-concentrate solution cleans and disinfects non-porous instruments and apparatus surfaces in a 10-minute period. UltraDose Germicide eliminates residual micro-organisms in a single step. It performs as an ultrasonic cleaner, disinfectant, deodorant, germicidal, fungicidal, viricidal, and tuberculocidal, all in one. UltraDose Germicide Solution works well on MRSA. This cleaning solution is designed for use in an ultrasonic cleaner.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.