Learn More
TARGETMOL CHEMICALS INC ULK-101 5MG
Supplier: TARGETMOL CHEMICALS INC T5403
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ULK-101 is a potent and selective ULK1 inhibitor ( IC50s 8.3/30 nM for ULK1/ULK2). It can suppress autophagy. purity: 99%