Learn More
Rockland Immunochemicals TURKEY RED BLOOD CELLS (RBC) 10% Washed Pooled Cells - R408-0050
SDP
Supplier:  Rockland Immunochemicals R4080050
Encompass_Preferred
Turkey whole blood is washed to remove the platelet rich plasma, buffy coat layer, and leukocytes (WBC). Red blood cells are supplied as a 10 percent suspension in phosphate buffered saline (PBS). Turkey red blood cells are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Turkey red blood cells are perishable and are collected and processed upon receipt of your order.
Catalog No. 50-105-9138
Explore available promotions