Learn More
Chemglass Life Sciences Tube Rotator, Analog, Single/Fixed Speed, 120V
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CLS3965100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Tube Rotator, Analog, Single/Fixed Speed, 120V. • “Plug & Play” operation with 360º rotation• Easy exchange of paddles, no tools needed• Quiet, high-quality, maintenance-free motorTube rotators are used in a variety of fields including molecular biology, biochemistry, histochemistry as well as in clinical applications. Interchangeable paddles accommodate different size tubes and can be assembled on a single rotisserie for simultaneous rotation of a variety of tubes. Individual paddle direction can be independently adjusted from 0-90º. To change the direction of rotation, just touch a paddle and the rotator will automatically reverse direction. Small enough to use on a crowded bench or in a hood.Each tube rotator includes a base unit, rotisserie and selection of paddles. Supplied with the following rotisserie paddles: (2) for 10/15mL tubes, (2) for 5/7mL tubes, (2) for 1.5/2mL tubes, (2) for 0.5/0.8mL tubes, and (1) for 50mL tubes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.