Learn More
Ace Glass, Inc. Tube, pressure, 210mL, 150psi, 38.1mm OD, 30.5cm long, complete with #15 back-seal plug
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 8648106
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
185mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 back-seal plug. Dimensions: 38.1mm O.D. x 30.5cm long.