Learn More
Ace Glass, Inc. Tube, pressure, 15mL, 150psi, 25.4mm OD, 10.2cm long, complete with #15 front-seal plunger valve with thermowell
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 8648164
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
15mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 front-seal plunger valve with Thermowell. Dimensions: 25.4mm O.D. x 10.2cm long.