Learn More
Electron Microscopy Sciences Tri-Pour Beaker Cap for 100 ml beaker
SDP
Supplier:  Electron Microscopy Sciences 60971
Encompass_Preferred
The caps are available for all Tri-Pour beakers. The caps are wax impregnated, have lift-tabs, and may be written on for sample identification.
Use: 100 ml
Catalog No. 50-189-8985
Explore available promotions