Learn More
Wilmad Labglass TRAP VAC 2PC 60X300MM COMP
Supplier: Wilmad Labglass LG11026110
Vacuum Trap, Two-Piece, with Hose Connectors, Complete. Body O.D.: 60mm; Body Length: 300mm; Joint: [ST] 55/50; Hose Connectors: 3/8 in.. For removal of vapors from a vacuum line by condensation with liquid air or dry ice slurry mixture
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov