Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - TU (T4 Uptake)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Supplier:  Tosoh Bioscience 0020370
 View more versions of this product
Catalog No. 21-279-51
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
  • These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
  • Saves hands-on time for operators and reduces waste
  • No pre-mixing or on-board refrigeration
  • Calibration stability lasts up to 90 days on most assays
Specifications
Specifications
TU Calibrator Set
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
12 x 1 mL