Learn More
Restek™ Topaz Straight Inlet Liner
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Features revolutionary technology and inertness to deliver next level True Blue Performance
$131.00 - $371.00
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Diameter (Metric) Outer | Length (Metric) | For Use With (Application) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Diameter (Metric) Outer | Length (Metric) | For Use With (Application) | Price | Quantity & Availability | ||
06-611-358
![]() |
Restek™
23460 |
0.53 mm | 4.6 mm | 54 mm | Bruker/Varian GCs |
Pack of 5 for $230.00
|
|
||
06-611-359
![]() |
Restek™
23461 |
0.80 mm | 4.6 mm | 54 mm | Bruker/Varian GCs |
Pack of 5 for $221.00
|
|
||
06-611-346
![]() |
Restek™
23448 |
1 mm | 6.3 mm | 78.5 mm | Thermo GCs |
Pack of 5 for $131.00
|
|
||
06-611-335
![]() |
Restek™
23436 |
1 mm | 2.75 mm | 120 mm | Thermo GCs |
Pack of 5 for $298.00
|
|
||
06-611-365
![]() |
Restek™
23471 |
1.5 mm | 3.5 mm | 95 mm | Shimadzu GCs |
Pack of 10 for $371.00
|
|
||
06-611-334
![]() |
Restek™
23435 |
1.6 mm | 4 mm | 95 mm | Shimadzu GCs |
Pack of 5 for $256.00
|
|
||
06-611-336
![]() |
Restek™
23437 |
2 mm | 2.75 mm | 120 mm | Thermo GCs |
Pack of 5 for $348.00
|
|
||
06-611-337
![]() |
Restek™
23439 |
3 mm | 8 mm | 105 mm | Thermo GCs |
Pack of 5 for $140.00
|
|
||
06-611-315
![]() |
Restek™
23318 |
3.5 mm | 5 mm | 95 mm | Shimadzu GCs |
Pack of 5 for $154.00
|
|
||
06-611-343
![]() |
Restek™
23445 |
4 mm | 6.3 mm | 78.5 mm | Thermo GCs |
Pack of 5 for $139.00
|
|
||
06-611-319
![]() |
Restek™
23323 |
5 mm | 8 mm | 105 mm | Thermo GCs |
Pack of 5 for $169.00
|
|
||
Description
- Deactivation: Unbelievably low breakdown for accurate and precise low-level GC analyses
- Reproducibility: Unbeatable manufacturing controls and QC testing for superior reliability across compound classes
- Productivity: Unparalleled cleanliness for maximized GC uptime and lab throughput
- 100% Satisfaction: If a liner does not perform to expectations, it will be replaced or account will be credited
- Patented
Specifications
Topaz |