Learn More
Miles Scientific TLC Sprayer Kit 3/pk
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Miles Scientific A4001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This is a refillable sprayer powered by its own self-contained replaceable pressure unit. It can be used with all common TLC reagents and delivers up to 16 oz of reagent from each pressure unit. Aspirating tube is made of polypropylene and the particulate filter is nylon. Meets EPA requirements for approved propellants (CFC free). Not for use with alcoholic sulfuric acid. Aqueous sulfuric acid is OK with immediate cleaning with water.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.