Learn More
Macherey-Nagel TLC ADSORBENT (BULK), SILICA G
Supplier: Macherey-Nagel 816320.5
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
TLC adsorbent (bulk), Silica gel G, contains gypsum, with UV indicator F254 5kg
This item is not returnable.
View return policy