Learn More
Apexbio Technology LLC Timolol Maleate, 100mg. Cas: 26921-17-5 MFCD: MFCD00058356. Non-selective, beta-adrenergic receptor antagonist
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Apexbio Technology LLC B1350100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
(S)-Timolol maleate, is a potent non-selective Beta-adrenergic receptor antagonist (Ki values are 1.97 and 2.0 nM for beta1 and Beta2 receptor subtypes respectively). 26921-17-5. MFCD00058356