Learn More
Sonoco™ ThermoSafe U-tek™ Gel Pack
Phase Change Material (PCM) gel pack for shipping temperature-sensitive materials.
Supplier: Sonoco™ 598
Description
- Gel will phase change at -1 deg. or -23 deg.
- Maintains critical temperatures of serums, test samples, live or frozen tissues, organs, and pharmaceuticals for extended periods.
- Laminated, metallized film exterior provides radiant heat barrier and resists punctures.
- Thermal solutions for temperature critical packages during transit
- condition coolant in freezer below -1 deg. or -23 deg. before shipments.
- For frozen or refrigerated shipments.
- Alternative to dry ice.
- Can be paired with our Mini Mailer(440).
- Available in several sizes
Specifications
8/Cs. | |
1 in. | |
-1°C | |
9.37 in. | |
Laminated, Metallized Film | |
907 g | |
19.5 cm |
U-tek Refrigerant Pack | |
Ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or Carbon dioxide | |
2.54 cm | |
23.7 cm | |
32 oz. | |
7.75 in. | |
9.37 x 7.75 x 1 in. (23.7 x 19.5 x 2.54 cm) |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov