Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Thermolite Plus Septa
Ultra-low bleed minimizes background signal
Supplier: Restek™ 23878
Description
- Usable to 350°C inlet temperature (except 17 mm inlets max temp 300°C)
- New plasma coating eliminates sticking in injection port
- Precision molding ensures consistent, accurate fit
- Septa have a CenterGuide design to minimize coring
- (Not available on Shimadzu plug, 6, 7, 8, 9.5, and 10 mm)
- 5 mm septa partially predrilled for improved puncturability
- Preconditioned and ready to use
- Packaged in ultra-clean blister packs
- Each batch GC-FID tested
Note:
- Due to differences in inlet design, actual septum temp for a given inlet set point can vary by manufacturer
- Recommended using only BTO septa in older Thermo TRACE and Focus GCs
- Does not apply for Thermo TRACE 1300/1310 GCs
- Septa can be used only to max recommended temp
Specifications
10 mm | |
Chromatography | |
50 Pk. |
Thermolite Plus Septa | |
350°C |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction