Learn More
MilliporeSigma™ Supelco™ Thermogreen™ LB-2 Septa, Pre-drilled
For use with SPME applications to reduce septum coring
Supplier: MilliporeSigma™ Supelco™ 23167
Description
For use with:
- Minimizing extraction onto or loss of analytes off the solid phase microextraction (SPME) fiber from the vial during analysis by GC-MS
- Avoiding carrier gas leaks, extraneous peaks, and phthalate contamination in GC instruments
- Use in conjunction with a thick needle protector to avoid damage to GC instruments irreversibly caused by microsyringes
- Influent sampling ports in gas chromatographs
Specifications
7/16 in. | |
SPME | |
Pre-drilled | |
Septa, LB-2 |
11 mm | |
25/Pk. | |
Thermogreen |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC