Learn More
The GLOBE Program™ Equipment
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Supplier: Bacharach Inc 127043
Description
GLOBE is a worldwide hands-on, primary and secondary school-based science and education program. The equipment specified here is based on descriptions and specifications contained in The GLOBE Program Teacher's Guide. Skill level recommendations are indicated as Elementary, Middle, and High for each piece of specified equipment.
- Sling Psychrometer measures relative humidity
- Slide rule calculator correlates wet and dry bulb thermometer indications for direct reading
- Built-in water reservoir
- -50°C to +50°C
- Red spirit filled
Specifications
Spirit Fill Sling Psychrometer | |
19 cm | |
Built-in water reservoir |
7.5 in. | |
Determines percent relative humidity |