Learn More
General Data Healthcare™ TFM™ Colored Tissue Freezing Medium
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Variety of colors to easily differentiate multiple specimens during sectioning and staining
Supplier: General Data Healthcare™ TFMY
Description
- Unique formula reduces curling, allowing easier pickup of flat serial sections
- Reduced water content minimizes freeze fractures
Order Info
Supplied in 4 oz. (118mL) plastic squeeze bottles. Complete set includes one of all five medium colors in a wooden rack.
Specifications
212°C | |
0°C | |
118mL |
Yellow | |
Tissue Freezing Medium | |
Embedding matrix for frozen sections |