Learn More
Biotium Tetrodotoxin, With Citrate, 1 mg
Supplier: Biotium 00061
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Tetrodotoxin (TTX) reversibly blocks excitable sodium channels and has been a widely used tool for studies of excitable membranes of nerve, skeletal muscle and cardiac muscle cells. Biotium offers high quality tetrodotoxin with or without citrate, at a competitive price. Tetrodotoxin, citrate-free can be dissolved in slightly acidic buffer. Tetrodotoxin, with citrate can be dissolved in water to obtain TTX in pH 4.8 citrate buffer.Properties: C11H17N3O8; MW: 319.28; [4368-28-9]
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.