Learn More
TARGETMOL CHEMICALS INC TENOFOVIR DISOPROXIL FUM 100MG
Supplier: TARGETMOL CHEMICALS INC T2409L
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tenofovir Disoproxil Fumarate (GS-1278 Disoproxil Fumarate) is a pro-drug fumaric acid salt form of tenofovir a nucleoside reverse transcriptase inhibitor analog of adenosine. Tenofovir disoproxil fumarate is prescribed to treat HIV and chronic hepatitis B virus (HBV) in adults. purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.