Learn More
IMEB, Inc The Tek-Select Lighted Water Bath
SDP
Supplier:  IMEB, Inc WB320
Encompass_Preferred
The Tek-Select lighted tissue flotation bath eliminates wrinkles and distortion of tissue sections for microscopy. A removable glass dish makes for a simple task of cleaning. The bath is a low profile design, with a microprocessor controller for heating. The temperature probe ensures accurate temperature control. Smart design, combines both water bath & slide dryer with separate temperature controls for water bath and slide dyer. Digital modular LED display shows set temperature, working temperature and other various parameters of working status.
Catalog No. NC2029578
Explore available promotions