Learn More
Electron Microscopy Sciences Technovit Polymerization Device (TPD)
Supplier: Electron Microscopy Sciences 1465322
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The unique EMS Technovit Polymerization Device (TPD) is a blue light curing device for all Technovit resins. The unit has built-in reflectors for homogenous curing of the blocks. The TPD fits up to 9 embedding molds and is generously sized.