Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ TCID
Selective ubiquitin C-terminal hydrolase-L3 (UCH-L3) inhibitor
$187.92 - $774.88
Specifications
Purity | >99% |
---|---|
Product Type | TCID |
Molecular Weight (g/mol) | 283.92 |
Inhibitors | TCID |
Description
Selective ubiquitin C-terminal hydrolase-L3 (UCH-L3) inhibitor (IC50 = 0.6 μM). Exhibits >100-fold selectivity for UCH-L3 over UCH-L1. Diminishes glycine transporter GlyT2 ubiquitination in brain stem and spinal cord primary neurons.Specifications
>99% | |
TCID | |
283.92 | |
TCID |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title