Learn More
TARGETMOL CHEMICALS INC TAZAROTENE 50MG
Supplier: TARGETMOL CHEMICALS INC T6696
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tazarotene (Zorac) is a synthetic topical retinoid. Tazarotene induces the expression of tazarotene-induced gene 3 (TIG3) a tumor suppressor gene. In psoriasis tazarotene normalizes abnormal keratinocyte differentiation and reduces their hyperproliferation. purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.