Learn More
Ampco Safety Tools T-Handle Wrench, Hex Key (Imperial)
AMPCO™ Hex Key T-Handle Wrench performs in high-risk and demanding environments.
$159.20 - $292.00
Specifications
Product Type | Hex Key T-handle |
---|---|
Material | Beryllium Copper |
Certifications/Compliance | Factory Mutual (FM) Approved |
Alert:
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.; WARNING: Cancer - www.P65Warnings.ca.gov
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-175-115
|
Ampco Safety Tools
WT1/4 |
1/4 in. |
Each for $225.50
|
|
|||||
19-175-116
|
Ampco Safety Tools
WT1/8 |
1/8 in. |
Each for $175.50
|
|
|||||
19-175-120
|
Ampco Safety Tools
WT3/16 |
3/16 in. |
Each for $194.90
|
|
|||||
19-175-121
|
Ampco Safety Tools
WT3/32 |
3/32 in. |
Each for $167.30
|
|
|||||
19-175-122
|
Ampco Safety Tools
WT3/8 |
3/8 in. |
Each for $292.00
|
|
|||||
19-175-125
|
Ampco Safety Tools
WT5/16 |
5/16 in. |
Each for $254.00
|
|
|||||
19-175-126
|
Ampco Safety Tools
WT5/32 |
5/32 in. |
Each for $175.50
|
|
|||||
19-175-127
|
Ampco Safety Tools
WT5/64 |
5/64 in. |
Each for $159.20
|
|
|||||
19-175-130
|
Ampco Safety Tools
WT7/32 |
7/32 in. |
Each for $191.80
|
|
|||||
Description
- Made of AMPCO invented, Factory Mutual approved alloy
- Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
- Non-Sparking
- Non-Magnetic
- Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Hex Key T-handle | |
Beryllium Copper | |
Factory Mutual (FM) Approved |
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.