Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUCHI SyncorePlus Analyst Parallel Evaporator System
![SDP](/content/dam/fishersci/glyphs/sdp.png)
SyncorePlus Analyst is specially designed to evaporate samples to a specified volume as a pre-analytical step, with an S-Condensor the final volume is 1 mL. Includes a V-300 vacuum pump with secondary condenser, I-300 Pro interface, and F-308 recirculating chiller.
Supplier: BUCHI 11SQA31S00121
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Description
- Safely and efficiently concentrate up to 12 samples to a predefined residual volume
- Residual volumes range from 0.3 mL, 1 mL, to 3 mL per sample
- Interchangeable racks and a wide range of accessories are available to create the ideal system to meet laboratory needs
- Individual sample sealing, and the use of inert materials inhibits cross-contamination
- A cooled appendix protects heat-sensitive analytes
Specifications
60 to 400 rpm | |
Sample Evaporation | |
115 V | |
SyncorePlus Analyst | |
EasyFill R-12 Analyst | |
520 x 500 x 325 mm |
12 x 125 mL | |
50/60 Hz | |
30 kg | |
20°C to 100°C | |
1250 W |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction