Learn More
Thermo Scientific Dionex AERS 500 Carbonate Anion Electrolytically Regenerated Suppressor for Carbonate Eluents (4 mm)
Supplier: Thermo Scientific 085029
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Get higher sensitivity and lower noise for your anion separations. The Thermo Scientific Dionex AERS 500 Carbonate Electrolytically Regenerated Suppressor is designed for anion exchange chromatography using carbonate-based eluents.
This item is not returnable.
View return policy
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)