Learn More
World Precision Instrument SU-MPC200 Motorized Micromanipulator Controllers-505093
Supplier: World Precision Instrument 505093
Neurobiological experiments are becoming more complex. Many require multiple manipulators with control units that quickly become space and/or cost prohibitive. The MPC-200 is the solution you have been asking for. A single controller capable of running 2 manipulators! With the simplicity of the SU-MP225 controller, it was expanded to run two manipulators from a single controller/ROE. The MPC-200 works with one or two of our world-renowned mechanicals, the SU-MP285 (SU-MPC385), the SU-MP225 (SU-MPC325) or the narrow format SU-MP265 (SU-MPC365).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.