Learn More
Foxx Life Sciences EZBio MTO Single Use (SUT) Media Bottle Assembly, 500mL, PETG, VersaCap 38-430, 2 Port (2x) 1/4 HB Adapter, Non-Sterile, 10/CS
SDP
Supplier:  Foxx Life Sciences 5379004FLS
Encompass_Preferred
Foxx Life Sciences EZBio Made To Order MTO 500mL PETG (Polyethylene Terephthalate Glycol-modified) Single Use Bottles. The EZBio MTO Single-Use Non Sterile PETG Media Bottle Assembly is an Innovative and Cost-Efficient Solution. Assemblies includes a PETG Bottle, Open-Cap, 2-Ported Adapter (Insert) with 2x 1/4" (6.4mm) Hose Barb Ports and one extra closed cap for storage. Bottle volumes range from 125ml to 2L. 125ml to 1000mL (1L) bottle assemblies come with an open-top 38-430 cap and two port adapter with 2x 1/4" (6.4mm) hose barbs. 2L Bottles come with an open top 53B VersaCap Closure and two port adapter with 2x 1/4" (6.4mm). Assemblies are made entirely with USP Class VI components and are perfect for Fluid Transfer and Storage Applications.
Catalog No. 50-206-0639
Explore available promotions