Learn More
Kyfora Bio Stigmasterol 1 GR
Supplier: Kyfora Bio 144971
Stigmasterol is a naturally occurring phytosterol lipid that is growing in the bioprocessing space for its ability to improve lipid nanoparticle (LNP)-based mRNA transfection efficiency. LNPs are well known for facilitating the safe and effective delivery of mRNA to target cells thus enabling therapeutic action. Cholesterol derivatives like stigmasterol offer two major benefits to LNP formulations: they can improve the stability of the LNP and they can increase the transfection efficiency in both in cell cultures and in vivo. Polysciences high purity transfection grade stigmasterol can be used in conjunction with our other cationic lipid products to provide reliable performance for a variety of bioprocessing applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.