Learn More
Sterilized Sample Ports for CTS™ Rotea™ Single-Use Kit
The Gibco CTS Rotea Single-Use Kit is part of the Gibco CTS Rotea Counterflow Centrifugation System. Each kit can perform a wide range of cell processing steps, making it ideal for wash & concentration, separation, isolation, and more.
Supplier:  THERMO SCI LSG 3P ATTUNE A50111
Catalog No. 50-272-3570
Explore available promotions
Add to Cart
Description
Description
  • The Gibco CTS Rotea Single-Use Kit is part of the Gibco CTS Rotea Counterflow Centrifugation System
  • Each kit can perform a wide range of cell processing steps, making it ideal for wash & concentration, separation, isolation, and more
  • The single-use kit provides flexibility for process development and easily supports commercial-scale manufacturing
  • Each kit is compatible with sterile luer connectors or thermal welding
  • These Sterilized Sample Ports for use with the kit enable collection of product samples during protocol execution, for process development
  • Sample port assembly consists of a needleless port joined with short lengths of PVC tube
  • PVC tube is sealed to enable thermal welding
  • Sample port assemblies and PVC tubing are gamma-irradiated and individually packaged
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 Pk.
No
CTS Rotea
Product Suggestions
Product Suggestions