Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Globe Scientific Test Tubes with Screw Cap
Multipurpose screw cap test tubes suit variety of applications.
Supplier: Globe Scientific 6156
Description
- Designed to be leak-tight allowing tubes to be used for storage or transport
- Screw caps are polyethylene
- Round bottom
Specifications
Red | |
Natural | |
120 mm | |
Attached screw cap | |
Sterile | |
No |
Polyethylene | |
15 mL | |
Polystyrene | |
150/Bag. | |
Not Autoclavable | |
Test Tube |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction