Learn More
Sigma Aldrich Fine Chemicals Biosciences Stand for Bubble Flowmeter | Heavy-Duty Lab Stand | Steel and Zinc-Plated | 1 Unit
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5094421EA
Encompass
STAND FOR BUBBLE FLOWMETERThe STAND FOR BUBBLE FLOWMETER is a support stand designed to hold laboratory equipment such as flowmeters. The stand features a steel base with a black enamel finish measuring 4 inches x 6 inches. A removable zinc-plated steel rod extends vertically from the base to provide a mounting point for equipment. The heavy-duty version includes an added steel plate on the back of the base for increased stability.Keywords: flowmeter stand, flowmeter support, laboratory stand, laboratory equipment stand, steel stand, zinc-plated stand, heavy-duty lab stand
Catalog No. NC2330909
Explore available promotions