Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stainless Steel wire, 0.51mm (0.02in) dia, Type 304, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 040946BZ

 View more versions of this product

Catalog No. AA40946BZ


Order in Bulk

Request bulk or custom formats

Add to Cart

Description

Description

It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stainless Steel wire, Type 304
Fe:Cr:Ni; 70:19:11 wt%
150 cm
Odorless
0.51mm (0.02 in.) dia.
MFCD00148499
Insoluble in water.
Wire
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350-H372

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

RUO – Research Use Only