Learn More
Stainless Steel wire, 0.51mm (0.02in) dia, Type 304, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 040946BW
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel wire, Type 304 | |
Fe:Cr:Ni; 70:19:11 wt% | |
50 cm | |
Odorless |
0.51mm (0.02 in.) dia. | |
MFCD00148499 | |
Insoluble in water. | |
Wire |
Safety and Handling
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only