Learn More
Stainless Steel powder, -140 mesh, Type 303-L, Thermo Scientific Chemicals
$248.65
Specifications
Chemical Name or Material | Stainless Steel powder, Type 303-L |
---|---|
Avg. Mol. Wt. or Mol. Wt. Range | Fe:Cr:Ni; 70:17:13 wt% |
MDL Number | MFCD00148499 |
Solubility Information | Insoluble in water. |
Mesh Size | −140 Mesh |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA11084A1
|
Thermo Scientific Chemicals
011084A1 |
1 kg |
Each of 1
for $248.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This offers moderate corrosion resistance. Alloy 303 is frequently used in applications that require parts to be heavily machined. It is used in nuts and bolts, aircraft fittings, gears and screws.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel powder, Type 303-L | |
MFCD00148499 | |
−140 Mesh |
Fe:Cr:Ni; 70:17:13 wt% | |
Insoluble in water. | |
Powder |
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.