Learn More
Stainless Steel powder, -100 mesh, Type 316-L, Thermo Scientific Chemicals
$87.00 - $249.00
Specifications
Chemical Name or Material | Stainless Steel powder, Type 316-L |
---|---|
Color | Gray |
Avg. Mol. Wt. or Mol. Wt. Range | Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% |
MDL Number | MFCD00148499 |
Solubility Information | Insoluble in water. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA1108936
|
Thermo Scientific Chemicals
01108936 |
500 g |
Each for $87.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AA11089A3
|
Thermo Scientific Chemicals
011089A3 |
2 kg |
Each for $249.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
It has an extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment. It finds its application in diamond tools, grinding and P/M parts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel powder, Type 316-L | |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
Insoluble in water. | |
Powder |
Gray | |
MFCD00148499 | |
−100 Mesh |
Safety and Handling
GHS H Statement
H372-H351-H319-H317-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.