Learn More
Stainless Steel foil, 0.1mm (0.004in) thick, Type 304, Thermo Scientific Chemicals
$27.65 - $51.25
Specifications
Chemical Name or Material | Stainless Steel |
---|---|
Color | Silver |
Appearance | Silver -colored foil |
Molecular Formula | Fe:Cr:Ni; 70:19:11 wt% |
MDL Number | MFCD00148499 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA41583FF
|
Thermo Scientific Chemicals
041583FF |
25 x 25 mm |
Each
for $27.65
Save 11% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA41583FI
|
Thermo Scientific Chemicals
041583FI |
50 x 50 mm |
Each
for $36.65
Save 11% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA41583GH
|
Thermo Scientific Chemicals
041583GH |
100 x 100 mm |
Each for $51.25
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel | |
Silver -colored foil | |
MFCD00148499 | |
Odorless |
Silver | |
Fe:Cr:Ni; 70:19:11 wt% | |
It is insoluble in water. | |
Foil |
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.